siRNA Details
| Gene Symbol: SLC22A18AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302862.1 | NP_001289791.1 | 3 | 919 | ||
| NM_007105.3 | NP_009036.2 | 4 | 1314 | |||
| XM_017017832.1 | XP_016873321.1 | 2 | 2352 | |||
| XM_017017833.1 | XP_016873322.1 | 4 | 2735 | |||
| XM_017017834.1 | XP_016873323.1 | 2 | 2016 | |||
| XM_017017835.1 | XP_016873324.1 | 2 | 1795 | |||
| XM_017017836.1 | XP_016873325.1 | 2 | 660 | |||
| GenBank mRNA | AF035407.1 | - | 2 | 681 | ||
| AF037066.1 | - | 4 | 1110 | |||
| BC030237.1 | - | 3 | 475 | 4185944 | ||

SLC22A18AS
Genomic Map