siRNA Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017372.2 | NP_001017372.1 | 2 | 1513 | ||
| NM_001317984.1 | NP_001304913.1 | 3 | 1337 | |||
| NM_014031.4 | NP_054750.1 | 3 | 1288 | |||
| GenBank mRNA | AF064254.1 | - | 2 | 655 | ||
| AK313456.1 | - | 3 | 936 | |||
| AL833117.1 | - | 2 | 1086 | |||
| BX537383.1 | - | 3 | 898 | |||
| CR457129.1 | - | 2 | 502 | |||

SLC27A6
Genomic Map