siRNA Details
| Gene Symbol: SLC39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271957.1 | NP_001258886.1 | 3 | 559 | ||
| NM_001271958.1 | NP_001258887.1 | 3 | 396 | |||
| NM_001271959.1 | NP_001258888.1 | 3 | 565 | |||
| NM_001271960.1 | NP_001258889.1 | 4 | 457 | |||
| NM_014437.4 | NP_055252.2 | 4 | 703 | |||
| GenBank mRNA | AF132942.1 | - | 2 | 253 | ||
| AF151829.1 | - | 3 | 274 | |||
| AJ243649.1 | - | 3 | 290 | |||
| AJ271671.1 | - | 3 | 304 | |||
| AK074943.1 | - | 3 | 513 | |||
| AK075257.1 | - | 3 | 327 | |||
| AK297983.1 | - | 4 | 271 | |||
| AK300244.1 | - | 4 | 263 | |||
| BC002563.2 | - | 3 | 278 | 3138575 | ||
| BC003152.2 | - | 2 | 250 | 2958651 | ||
| BC007886.2 | - | 3 | 275 | 4122086 | ||
| BC014303.2 | - | 4 | 683 | 3942586 | ||
| BG704399.1 | - | 3 | 567 | |||
| CR457217.1 | - | 2 | 217 | |||
| DA052424.1 | - | 3 | 397 | |||
| DA354647.1 | - | 3 | 541 | |||
| DA711655.1 | - | 3 | 406 | |||
| DA818910.1 | - | 3 | 383 | |||
| DB001496.1 | - | 4 | 457 | |||

SLC39A1
Genomic Map