siRNA Details
| Gene Symbol: OAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032731.1 | NP_001027903.1 | 2 | 431 | ||
| NM_002535.2 | NP_002526.2 | 2 | 431 | |||
| NM_016817.2 | NP_058197.2 | 2 | 431 | |||
| GenBank mRNA | AK292796.1 | - | 2 | 320 | ||
| AK292906.1 | - | 2 | 348 | |||
| BC010625.1 | - | 2 | 417 | |||
| BC023637.2 | - | 2 | 352 | |||
| BC049215.1 | - | 2 | 332 | 5517804 | ||
| BC053870.1 | - | 2 | 354 | |||
| BC069052.1 | - | 2 | 343 | |||
| BE885222.1 | - | 488 | ||||
| M87284.1 | - | 2 | 310 | |||
| M87434.1 | - | 2 | 310 | |||

OAS2
Genomic Map