siRNA Details
| Gene Symbol: MRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317947.1 | NP_001304876.1 | 3 | 698 | ||
| NM_018146.3 | NP_060616.1 | 4 | 943 | |||
| GenBank mRNA | AF177344.2 | - | 4 | 999 | ||
| AK001443.1 | - | 4 | 919 | |||
| AK222900.1 | - | 4 | 919 | |||
| AK225128.1 | - | 4 | 900 | |||
| BC011550.1 | - | 4 | 905 | 4620063 | ||
| BC050614.1 | - | 4 | 901 | 5248052 | ||

MRM3
Genomic Map