siRNA Details
| Gene Symbol: SLC5A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305005.2 | NP_001291934.1 | 9 | 2562 | ||
| NM_001305006.2 | NP_001291935.1 | 8 | 2336 | |||
| NM_001305007.2 | NP_001291936.1 | 9 | 2528 | |||
| NM_021815.4 | NP_068587.1 | 9 | 2565 | |||
| XM_011511580.2 | XP_011509882.1 | 7 | 2116 | |||
| XM_017004628.1 | XP_016860117.1 | 7 | 3336 | |||
| XM_017004629.1 | XP_016860118.1 | 7 | 5844 | |||
| GenBank mRNA | AB043997.1 | - | 9 | 2540 | ||
| BC111524.1 | - | 9 | 2470 | 40034612 | ||
| BC111525.1 | - | 9 | 2512 | 40034618 | ||

SLC5A7
Genomic Map