siRNA Details
| Gene Symbol: SLC2A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030777.3 | NP_110404.1 | 2 | 1191 | ||
| XM_011529060.2 | XP_011527362.1 | 2 | 1274 | |||
| XM_011529061.2 | XP_011527363.1 | 2 | 2831 | |||
| XM_011529062.2 | XP_011527364.1 | 2 | 1285 | |||
| XM_011529063.2 | XP_011527365.1 | 2 | 1284 | |||
| XM_011529064.2 | XP_011527366.1 | 2 | 1286 | |||
| XM_011529065.2 | XP_011527367.1 | 2 | 1281 | |||
| XM_017028087.1 | XP_016883576.1 | 2 | 1184 | |||
| GenBank mRNA | AF248053.1 | - | 2 | 1344 | ||
| AF321240.1 | - | 2 | 1146 | |||
| AK055548.1 | - | 2 | 1191 | |||
| AK290961.1 | - | 2 | 1182 | |||
| AK315269.1 | - | 2 | 1182 | |||
| AL137188.3 | - | 1 | 1102 | |||
| BC101657.1 | - | 2 | 1190 | 8069163 | ||
| BC113423.1 | - | 2 | 1190 | 8322475 | ||

SLC2A10
Genomic Map