siRNA Details
| Gene Symbol: SLC2A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030777.3 | NP_110404.1 | 5 | 2762 | ||
| XM_011529060.2 | XP_011527362.1 | 5 | 2845 | |||
| XM_011529061.2 | XP_011527363.1 | 5 | 4402 | |||
| XM_011529062.2 | XP_011527364.1 | 4 | 2769 | |||
| XM_011529065.2 | XP_011527367.1 | 4 | 2716 | |||
| XM_017028087.1 | XP_016883576.1 | 4 | 2619 | |||
| GenBank mRNA | AF248053.1 | - | 5 | 2914 | ||
| AF321240.1 | - | 5 | 2717 | |||
| AK055548.1 | - | 5 | 2762 | |||
| AL137188.3 | - | 4 | 2673 | |||

SLC2A10
Genomic Map