siRNA Details
| Gene Symbol: SLC22A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080866.2 | NP_543142.2 | 10 | 1928 | ||
| XM_017017159.1 | XP_016872648.1 | 8 | 2533 | |||
| XM_017017160.1 | XP_016872649.1 | 9 | 1505 | |||
| GenBank mRNA | AB062418.1 | - | 10 | 1761 | ||
| AB074812.1 | - | 10 | 1909 | |||
| AF440402.1 | - | 10 | 1896 | |||
| AJ295270.1 | - | 10 | 1720 | |||
| AK074246.1 | - | 10 | 1925 | |||
| BC022379.1 | - | 7 | 1460 | |||
| BC126288.1 | - | 10 | 1810 | 8992004 | ||

SLC22A9
Genomic Map