siRNA Details
| Gene Symbol: AIFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018060.2 | NP_001018070.1 | 20 | 2024 | ||
| NM_001146288.1 | NP_001139760.1 | 20 | 2109 | |||
| NM_144704.2 | NP_653305.1 | 21 | 2045 | |||
| NR_027464.1 | 19 | 1981 | ||||
| GenBank mRNA | AK055035.1 | - | 21 | 2029 | ||
| AK094844.1 | - | 20 | 2013 | |||
| AK126917.1 | - | 4 | 2535 | |||
| AK295581.1 | - | 19 | 1884 | |||
| AK316042.1 | - | 19 | 1877 | |||
| BC032485.1 | - | 20 | 1991 | 5221203 | ||
| CR456342.1 | - | 20 | 1864 | |||

AIFM3
Genomic Map