siRNA Details
| Gene Symbol: SLC2A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145176.2 | NP_660159.1 | 2 | 1367 | ||
| XM_006715349.3 | XP_006715412.1 | 2 | 1384 | |||
| XM_017010311.1 | XP_016865800.1 | 2 | 1384 | |||
| GenBank mRNA | AK056554.1 | - | 1 | 106 | ||
| AK122628.1 | - | 2 | 1359 | |||
| AK226098.1 | - | 2 | 1238 | |||
| AL833602.1 | - | 2 | 1351 | |||
| AY046419.1 | - | 2 | 1329 | |||
| BC070149.1 | - | 2 | 1301 | 30371087 | ||

SLC2A12
Genomic Map