siRNA Details
| Gene Symbol: SLC5A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042450.2 | NP_001035915.1 | 12 | 1356 | ||
| NM_001270648.1 | NP_001257577.1 | 11 | 1275 | |||
| NM_001270649.1 | NP_001257578.1 | 11 | 1248 | |||
| NM_001282417.1 | NP_001269346.1 | 12 | 1465 | |||
| NM_152351.4 | NP_689564.3 | 12 | 1404 | |||
| XM_011523652.1 | XP_011521954.1 | 13 | 1623 | |||
| XM_011523653.2 | XP_011521955.1 | 12 | 1595 | |||
| XM_017024191.1 | XP_016879680.1 | 13 | 1672 | |||
| GenBank mRNA | AK057946.1 | - | 12 | 1405 | ||
| AK298345.1 | - | 11 | 1234 | |||
| BC034380.1 | - | 12 | 1301 | |||
| BC039868.1 | - | 12 | 1482 | 4157703 | ||
| BC062617.1 | - | 11 | 1229 | |||

SLC5A10
Genomic Map