siRNA Details
| Gene Symbol: SLCO6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289002.1 | NP_001275931.1 | 1 | 324 | ||
| NM_001289004.1 | NP_001275933.1 | 1 | 324 | |||
| NM_001308014.1 | NP_001294943.1 | 1 | 324 | |||
| NM_173488.4 | NP_775759.3 | 1 | 324 | |||
| XM_005271874.3 | XP_005271931.1 | 1 | 3521 | |||
| XM_011543147.2 | XP_011541449.1 | 1 | 3521 | |||
| XM_011543148.2 | XP_011541450.1 | 1 | 3521 | |||
| XM_011543154.2 | XP_011541456.1 | 1 | 3521 | |||
| GenBank mRNA | AF505657.1 | - | 1 | 295 | ||
| AK131445.1 | - | 1 | 221 | |||
| AY273897.1 | - | 1 | 287 | |||
| BC034976.2 | - | 1 | 326 | 4820512 | ||
| BG772379.1 | - | 1 | 314 | |||
| BM554439.1 | - | 1 | 311 | |||
| HY036202.1 | - | 1 | 326 | |||

SLCO6A1
Genomic Map