siRNA Details
| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143838.2 | NP_001137310.1 | 11 | 3116 | ||
| NM_001284509.1 | NP_001271438.1 | 12 | 3203 | |||
| NM_001284510.1 | NP_001271439.1 | 11 | 3125 | |||
| NM_177550.4 | NP_808218.1 | 12 | 3254 | |||
| GenBank mRNA | AK124317.1 | - | 1 | 4609 | ||
| AK127797.1 | - | 12 | 3152 | |||
| AK172779.1 | - | 5 | 2108 | |||
| AK172785.1 | - | 12 | 3200 | |||
| AY151833.1 | - | 12 | 3179 | |||
| BX647354.1 | - | 1 | 5360 | |||
| CR749584.1 | - | 7 | 2369 | |||

SLC13A5
Genomic Map