siRNA Details
| Gene Symbol: SLC37A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287498.1 | NP_001274427.1 | 4 | 419 | ||
| NM_032295.3 | NP_115671.1 | 4 | 419 | |||
| NM_207113.2 | NP_996996.1 | 4 | 419 | |||
| XM_011516626.2 | XP_011514928.1 | 6 | 916 | |||
| XM_011516627.2 | XP_011514929.1 | 6 | 915 | |||
| XM_017012712.1 | XP_016868201.1 | 6 | 892 | |||
| XM_017012713.1 | XP_016868202.1 | 6 | 911 | |||
| XM_017012714.1 | XP_016868203.1 | 6 | 932 | |||
| XM_017012715.1 | XP_016868204.1 | 6 | 935 | |||
| XM_017012716.1 | XP_016868205.1 | 6 | 939 | |||
| XM_017012717.1 | XP_016868206.1 | 6 | 911 | |||
| XM_017012718.1 | XP_016868207.1 | 6 | 910 | |||
| GenBank mRNA | AK074823.1 | - | 4 | 380 | ||
| AK296543.1 | - | 4 | 373 | |||
| AK299276.1 | - | 4 | 363 | |||
| AK300265.1 | - | 4 | 377 | |||
| AL136583.1 | - | 4 | 326 | |||
| BC028380.1 | - | 4 | 369 | 4811814 | ||
| BC046567.1 | - | 4 | 371 | 5246993 | ||
| CD101739.1 | - | 4 | 427 | |||
| DB481623.1 | - | 3 | 310 | |||

SLC37A3
Genomic Map