siRNA Details
| Gene Symbol: SLC37A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287498.1 | NP_001274427.1 | 14 | 1963 | ||
| NM_032295.3 | NP_115671.1 | 12 | 1709 | |||
| NM_207113.2 | NP_996996.1 | 15 | 2011 | |||
| XM_011516626.2 | XP_011514928.1 | 16 | 2356 | |||
| XM_011516627.2 | XP_011514929.1 | 15 | 2307 | |||
| XM_017012712.1 | XP_016868201.1 | 17 | 2484 | |||
| XM_017012713.1 | XP_016868202.1 | 14 | 2201 | |||
| XM_017012714.1 | XP_016868203.1 | 15 | 2281 | |||
| XM_017012715.1 | XP_016868204.1 | 15 | 2263 | |||
| XM_017012716.1 | XP_016868205.1 | 12 | 1965 | |||
| XM_017012717.1 | XP_016868206.1 | 13 | 2059 | |||
| GenBank mRNA | AK074823.1 | - | 15 | 1972 | ||
| AK126589.1 | - | 3 | 1155 | |||
| AL136583.1 | - | 12 | 1616 | |||
| BC028380.1 | - | 15 | 1961 | 4811814 | ||
| BC046567.1 | - | 14 | 1915 | 5246993 | ||

SLC37A3
Genomic Map