siRNA Details
| Gene Symbol: POLR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007055.3 | NP_008986.2 | 2, 3 | 312 | ||
| GenBank mRNA | AF021351.1 | - | 3, 4 | 179 | ||
| AK300329.1 | - | 2, 3 | 292 | |||
| AY091459.1 | - | 2, 3 | 179 | |||
| BC041089.1 | - | 2, 3 | 273 | 3915501 | ||
| DA103416.1 | - | 2, 3 | 312 | |||

POLR3A
Genomic Map