siRNA Details
| Gene Symbol: POLR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007055.3 | NP_008986.2 | 20 | 2816 | ||
| GenBank mRNA | AF021351.1 | - | 21 | 2683 | ||
| AK026023.1 | - | 11 | 1390 | |||
| AK300329.1 | - | 20 | 2796 | |||
| AY091461.1 | - | 5 | 490 | |||
| BC041089.1 | - | 20 | 2777 | 3915501 | ||

POLR3A
Genomic Map