siRNA Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290225.1 | NP_001277154.1 | 6 | 917 | ||
| NM_014754.2 | NP_055569.1 | 8 | 1087 | |||
| GenBank mRNA | AK293036.1 | - | 8 | 1071 | ||
| AK293513.1 | - | 4 | 397 | |||
| BC002376.2 | - | 6 | 890 | 2961075 | ||
| BC004192.2 | - | 8 | 1037 | 3509418 | ||
| BC004390.1 | - | 8 | 1037 | 3634879 | ||
| BC004502.2 | - | 4 | 508 | |||
| D14694.1 | - | 8 | 1034 | |||

PTDSS1
Genomic Map