siRNA Details
Gene Symbol: ACSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024560.2 | NP_078836.1 | 12 | 1659 | ||
XM_005269150.2 | XP_005269207.1 | 12 | 1593 | |||
XM_011538740.2 | XP_011537042.1 | 12 | 1308 | |||
GenBank mRNA | AK025616.1 | - | 12 | 1610 | ||
AK074267.1 | - | 2 | 83 | |||
AK074938.1 | - | 7 | 840 | |||
AX764480.1 | - | 12 | 1809 | |||
AX764482.1 | - | 12 | 1565 | |||
BC009317.2 | - | 12 | 1659 | 4120563 | ||
BC015769.1 | - | 12 | 1659 | 4902942 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
