siRNA Details
Gene Symbol: PTDSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030783.1 | NP_110410.1 | 3 | 518 | ||
XM_005253153.1 | XP_005253210.1 | 3 | 451 | |||
XM_011520391.2 | XP_011518693.1 | 3 | 1088 | |||
XM_011520392.2 | XP_011518694.1 | 4 | 515 | |||
XM_017018368.1 | XP_016873857.1 | 5 | 2910 | |||
XM_017018369.1 | XP_016873858.1 | 4 | 640 | |||
GenBank mRNA | AK311723.1 | - | 3 | 450 | ||
AL834357.1 | - | 3 | 432 | |||
BC001210.1 | - | 3 | 518 | 3503123 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
