siRNA Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304458.1 | NP_001291387.1 | 2 | 607 | ||
| NM_138364.3 | NP_612373.2 | 3 | 756 | |||
| XM_017008823.1 | XP_016864312.1 | 3 | 532 | |||
| XM_017008824.1 | XP_016864313.1 | 3 | 532 | |||
| XM_017008825.1 | XP_016864314.1 | 2 | 518 | |||
| XM_017008826.1 | XP_016864315.1 | 2 | 481 | |||
| GenBank mRNA | AK096703.1 | - | 2 | 502 | ||
| AK292904.1 | - | 3 | 689 | |||
| BC021250.2 | - | 3 | 511 | |||
| BC064403.1 | - | 3 | 655 | 5547611 | ||

PRMT9
Genomic Map