siRNA Details
| Gene Symbol: NDUFS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002496.3 | NP_002487.1 | 7 | 737 | ||
| XM_005274013.1 | XP_005274070.1 | 7 | 834 | |||
| XM_005274014.2 | XP_005274071.1 | 7 | 861 | |||
| XM_011545053.2 | XP_011543355.1 | 7 | 772 | |||
| GenBank mRNA | AK223114.1 | - | 7 | 727 | ||
| AK314546.1 | - | 7 | 662 | |||
| BC119753.1 | - | 3 | 294 | |||
| BC119754.2 | - | 7 | 666 | 40119290 | ||
| U65579.1 | - | 7 | 660 | |||

NDUFS8
Genomic Map