siRNA Details
| Gene Symbol: MAPK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001315.2 | NP_001306.1 | 2 | 651 | ||
| NM_139012.2 | NP_620581.1 | 2 | 651 | |||
| NM_139013.2 | NP_620582.1 | 2 | 651 | |||
| NM_139014.2 | NP_620583.1 | 2 | 651 | |||
| XM_006714998.3 | XP_006715061.1 | 2 | 126 | |||
| XM_011514310.2 | XP_011512612.2 | 2 | 919 | |||
| XM_017010299.1 | XP_016865788.1 | 2 | 693 | |||
| XM_017010300.1 | XP_016865789.1 | 2 | 693 | |||
| XM_017010301.1 | XP_016865790.1 | 2 | 693 | |||
| XM_017010302.1 | XP_016865791.1 | 2 | 693 | |||
| XM_017010303.1 | XP_016865792.1 | 2 | 693 | |||
| XM_017010304.1 | XP_016865793.1 | 2 | 693 | |||
| GenBank mRNA | AB074150.1 | - | 2 | 170 | ||
| AF100544.1 | - | 2 | 185 | |||
| AK291709.1 | - | 2 | 546 | |||
| AK303414.1 | - | 3 | 215 | |||
| BC000092.1 | - | 2 | 508 | 3505234 | ||
| BC031574.1 | - | 2 | 507 | 5181064 | ||
| BT006933.1 | - | 2 | 170 | |||
| CR536505.1 | - | 2 | 170 | |||
| FJ032367.1 | - | 2 | 170 | |||
| FJ032368.1 | - | 2 | 170 | |||
| L35263.1 | - | 2 | 532 | |||
| L35264.1 | - | 2 | 378 | |||

MAPK14
Genomic Map