siRNA Details
| Gene Symbol: LTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002344.5 | NP_002335.2 | 11 | 1584 | ||
| NM_206961.3 | NP_996844.1 | 10 | 1401 | |||
| XM_011521556.2 | XP_011519858.1 | 11 | 1632 | |||
| XM_011521557.2 | XP_011519859.1 | 10 | 1464 | |||
| XM_017022181.1 | XP_016877670.1 | 11 | 1647 | |||
| XM_017022182.1 | XP_016877671.1 | 10 | 1503 | |||
| GenBank mRNA | AK314006.1 | - | 11 | 1551 | ||
| BC045607.1 | - | 10 | 1828 | 5270700 | ||
| BC143903.1 | - | 5 | 587 | |||
| D16105.1 | - | 11 | 1503 | |||
| X52213.1 | - | 4 | 467 | |||
| X60702.1 | - | 10 | 1393 | |||

LTK
Genomic Map