siRNA Details
| Gene Symbol: DNALI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003462.3 | NP_003453.2 | 6 | 850 | ||
| XM_005271172.2 | XP_005271229.1 | 5 | 662 | |||
| GenBank mRNA | AF006386.1 | - | 6 | 827 | ||
| AK126963.1 | - | 2 | 1741 | |||
| AK225935.1 | - | 5 | 2889 | |||
| AK290502.1 | - | 6 | 774 | |||
| AK295199.1 | - | 5 | 593 | |||
| BC039074.1 | - | 6 | 833 | 5179639 | ||
| BC046117.1 | - | 6 | 800 | 6192652 | ||
| BX647913.1 | - | 6 | 850 | |||

DNALI1
Genomic Map