siRNA Details
| Gene Symbol: DNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281428.1 | NP_001268357.1 | 13 | 1395 | ||
| NM_012144.3 | NP_036276.1 | 13 | 1383 | |||
| XM_006716758.3 | XP_006716821.1 | 12 | 1026 | |||
| XM_011517846.1 | XP_011516148.1 | 13 | 1395 | |||
| XM_011517847.2 | XP_011516149.1 | 13 | 1395 | |||
| XM_011517848.1 | XP_011516150.1 | 13 | 1395 | |||
| XM_011517849.1 | XP_011516151.1 | 13 | 1395 | |||
| XM_011517850.2 | XP_011516152.1 | 13 | 1395 | |||
| XM_017014625.1 | XP_016870114.1 | 13 | 1383 | |||
| GenBank mRNA | AF091619.1 | - | 13 | 1300 | ||
| AK290696.1 | - | 13 | 1307 | |||
| AK302186.1 | - | 10 | 926 | |||
| AK303937.1 | - | 10 | 880 | |||
| BC030583.2 | - | 13 | 1311 | 4837991 | ||

DNAI1
Genomic Map