siRNA Details
| Gene Symbol: DNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172810.1 | NP_001166281.1 | 11, 12 | 1561 | ||
| NM_023036.4 | NP_075462.3 | 11, 12 | 1600 | |||
| XM_011525125.2 | XP_011523427.1 | 11, 12 | 1617 | |||
| GenBank mRNA | AF250288.1 | - | 11, 12 | 1557 | ||
| AJ295276.1 | - | 10, 11 | 1796 | |||
| AL137526.1 | - | 3, 4 | 543 | |||
| BC033013.1 | - | 10, 11 | 1474 | 4826432 | ||
| BC039582.1 | - | 11, 12 | 1551 | 5752153 | ||

DNAI2
Genomic Map