siRNA Details
| Gene Symbol: MYO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012334.2 | NP_036466.2 | 3 | 616 | ||
| XM_006714475.2 | XP_006714538.1 | 3 | 392 | |||
| GenBank mRNA | AB018342.2 | - | 3 | 603 | ||
| AF132021.1 | - | 3 | 228 | |||
| AF234532.1 | - | 3 | 370 | |||
| AF247457.2 | - | 3 | 301 | |||
| AK125246.1 | - | 3 | 600 | |||
| BC014956.1 | - | 3 | 612 | |||
| BC041694.1 | - | 3 | 588 | |||
| BC050682.1 | - | 3 | 522 | |||
| BC071965.1 | - | 2 | 171 | |||
| BC094838.1 | - | 2 | 171 | |||
| BC108736.1 | - | 3 | 591 | |||
| BC137168.1 | - | 3 | 325 | 9021165 | ||
| BC150285.1 | - | 3 | 603 | 8860110 | ||

MYO10
Genomic Map