siRNA Details
| Gene Symbol: MYO15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016239.3 | NP_057323.3 | 40 | 8068 | ||
| XM_017024714.1 | XP_016880203.1 | 37 | 8368 | |||
| XM_017024715.1 | XP_016880204.1 | 38 | 8431 | |||
| GenBank mRNA | AF144094.1 | - | 40 | 8068 | ||

MYO15A
Genomic Map