siRNA Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021019.4 | NP_066299.2 | 4 | 403 | ||
| NM_079423.3 | NP_524147.2 | 4 | 403 | |||
| GenBank mRNA | AB046613.1 | - | 4 | 302 | ||
| AK301198.1 | - | 3 | 581 | |||
| BC006781.2 | - | 2 | 233 | |||
| BC017455.1 | - | 4 | 290 | 3538373 | ||
| BC071661.1 | - | 4 | 286 | 5561575 | ||
| BC093066.1 | - | 4 | 299 | 30722094 | ||
| BG283793.1 | - | 4 | 402 | |||
| CN401857.1 | - | 4 | 287 | |||
| CR456839.1 | - | 2 | 157 | |||
| CR457014.1 | - | 3 | 262 | |||
| M22918.1 | - | 4 | 293 | |||
| M22920.1 | - | 3 | 271 | |||
| M31212.1 | - | 4 | 302 | |||
| U02629.1 | - | 4 | 300 | |||

MYL6
Genomic Map