siRNA Details
| Gene Symbol: KIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015074.3 | NP_055889.2 | 2, 3 | 364 | ||
| NM_183416.3 | NP_904325.2 | 2, 3 | 364 | |||
| GenBank mRNA | AB011163.2 | - | 2, 3 | 313 | ||
| AB017133.1 | - | 1, 2 | 196 | |||
| AB040881.2 | - | 2, 3 | 423 | |||
| AB088210.1 | - | 1, 2 | 112 | |||
| AB088211.1 | - | 1, 2 | 112 | |||
| AB088212.1 | - | 1, 2 | 112 | |||
| AB088213.1 | - | 1, 2 | 112 | |||
| AF257176.1 | - | 2, 3 | 265 | |||
| AK308121.1 | - | 2, 3 | 364 | |||
| AY043362.1 | - | 1, 2 | 116 | |||
| AY139835.1 | - | 2, 3 | 265 | |||
| BC115395.2 | - | 1, 2 | 200 | 40067993 | ||
| BF940695.1 | - | 2, 3 | 297 | |||

KIF1B
Genomic Map