siRNA Details
Gene Symbol: KLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134775.1 | NP_001128247.1 | 3 | 656 | ||
NM_001134776.1 | NP_001128248.1 | 3 | 535 | |||
NM_001318734.1 | NP_001305663.1 | 3 | 593 | |||
NM_022822.2 | NP_073733.1 | 3 | 630 | |||
XM_005274183.1 | XP_005274240.1 | 3 | 572 | |||
XM_005274184.1 | XP_005274241.1 | 3 | 593 | |||
GenBank mRNA | AK022449.1 | - | 3 | 522 | ||
AK022907.1 | - | 3 | 575 | |||
AK126489.1 | - | 1 | 1014 | |||
AK315725.1 | - | 3 | 525 | |||
AL136864.1 | - | 3 | 556 | |||
BC034373.1 | - | 3 | 518 | 4342879 | ||
DA198727.1 | - | 3 | 478 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
