siRNA Details
| Gene Symbol: KIF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198525.2 | NP_940927.2 | 9 | 2080 | ||
| XM_011521531.2 | XP_011519833.1 | 9 | 2190 | |||
| GenBank mRNA | AY358384.1 | - | 5 | 649 | ||
| BC104044.1 | - | 5 | 622 | 40026959 | ||
| BC112271.1 | - | 4 | 513 | 8327739 | ||
| BC112273.1 | - | 4 | 513 | 8327741 | ||
| BC143461.1 | - | 4 | 512 | 9051969 | ||

KIF7
Genomic Map