siRNA Details
| Gene Symbol: PCCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000282.3 | NP_000273.2 | 7 | 673 | ||
| NM_001127692.2 | NP_001121164.1 | 6 | 595 | |||
| NM_001178004.1 | NP_001171475.1 | 7 | 673 | |||
| XM_005254059.2 | XP_005254116.1 | 7 | 668 | |||
| XM_011521093.1 | XP_011519395.1 | 7 | 668 | |||
| XM_017020605.1 | XP_016876094.1 | 7 | 668 | |||
| XM_017020606.1 | XP_016876095.1 | 6 | 519 | |||
| XM_017020607.1 | XP_016876096.1 | 7 | 484 | |||
| XM_017020608.1 | XP_016876097.1 | 7 | 668 | |||
| XM_017020609.1 | XP_016876098.1 | 7 | 581 | |||
| XM_017020610.1 | XP_016876099.1 | 5 | 935 | |||
| XM_017020611.1 | XP_016876100.1 | 7 | 668 | |||
| XM_017020612.1 | XP_016876101.1 | 7 | 668 | |||
| XM_017020613.1 | XP_016876102.1 | 7 | 668 | |||
| XM_017020614.1 | XP_016876103.1 | 6 | 519 | |||
| XM_017020615.1 | XP_016876104.1 | 7 | 668 | |||
| XM_017020616.1 | XP_016876105.1 | 7 | 668 | |||
| XM_017020617.1 | XP_016876106.1 | 4 | 331 | |||
| XM_017020618.1 | XP_016876107.1 | 4 | 331 | |||
| GenBank mRNA | AF385926.1 | - | 7 | 605 | ||
| AK225288.1 | - | 7 | 573 | |||
| AK225299.1 | - | 7 | 571 | |||
| AK296771.1 | - | 6 | 493 | |||
| AK298318.1 | - | 7 | 595 | |||
| AK315503.1 | - | 7 | 571 | |||
| BC000140.2 | - | 7 | 545 | 3048998 | ||
| DA781564.1 | - | 6 | 493 | |||
| X14608.1 | - | 7 | 540 | |||
| Z36837.1 | - | 3 | 267 | |||

PCCA
Genomic Map