siRNA Details
| Gene Symbol: CFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145252.1 | NP_001138724.1 | 9 | 1535 | ||
| NM_002621.2 | NP_002612.1 | 10 | 1651 | |||
| XM_017029575.1 | XP_016885064.1 | 8 | 2157 | |||
| GenBank mRNA | BC015756.1 | - | 9 | 1469 | 4850121 | |
| M83652.1 | - | 9 | 1481 | |||
| X57748.1 | - | 9 | 1474 | |||

CFP
Genomic Map