siRNA Details
| Gene Symbol: PGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166424.1 | NP_001159896.1 | 2 | 206 | ||
| NM_002630.3 | NP_002621.1 | 2 | 206 | |||
| GenBank mRNA | AK223274.1 | - | 2 | 202 | ||
| AK301298.1 | - | 2 | 202 | |||
| AK301325.1 | - | 2 | 202 | |||
| BC042578.1 | - | 2 | 188 | |||
| BC073740.1 | - | 2 | 188 | 6296399 | ||
| J04443.1 | - | 2 | 146 | |||
| U75272.1 | - | 2 | 189 | |||

PGC
Genomic Map