siRNA Details
| Gene Symbol: HDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306146.1 | NP_001293075.1 | 11 | 2499 | ||
| NM_002112.3 | NP_002103.2 | 12 | 2598 | |||
| XM_017022094.1 | XP_016877583.1 | 12 | 2467 | |||
| XM_017022095.1 | XP_016877584.1 | 11 | 2368 | |||
| XM_017022096.1 | XP_016877585.1 | 11 | 2252 | |||
| XM_017022097.1 | XP_016877586.1 | 10 | 2550 | |||
| XM_017022098.1 | XP_016877587.1 | 9 | 2070 | |||
| GenBank mRNA | AK027221.1 | - | 5 | 2001 | ||
| M60445.1 | - | 12 | 2307 | |||
| X54297.1 | - | 12 | 2357 | |||

HDC
Genomic Map