siRNA Details
| Gene Symbol: PGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304451.1 | NP_001291380.1 | 3 | 272 | ||
| NM_001304452.1 | NP_001291381.1 | 3 | 351 | |||
| NM_002631.3 | NP_002622.2 | 3 | 272 | |||
| GenBank mRNA | AK290404.1 | - | 3 | 197 | ||
| AK298830.1 | - | 3 | 351 | |||
| AK300952.1 | - | 3 | 230 | |||
| AK303898.1 | - | 3 | 197 | |||
| AK304423.1 | - | 3 | 195 | |||
| BC000368.2 | - | 3 | 183 | 2819330 | ||
| CD700605.1 | - | 3 | 274 | |||
| HY049516.1 | - | 3 | 273 | |||
| U30255.1 | - | 2 | 152 | |||

PGD
Genomic Map