siRNA Details
| Gene Symbol: CDKL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037343.1 | NP_001032420.1 | 6 | 424 | ||
| NM_001323289.1 | NP_001310218.1 | 4 | 337 | |||
| NM_003159.2 | NP_003150.1 | 5 | 428 | |||
| GenBank mRNA | AK307444.1 | - | 5 | 425 | ||
| AL704691.1 | - | 6 | 463 | |||
| AY217744.1 | - | 5 | 396 | |||
| HQ171445.1 | - | 4 | 175 | |||
| Y15057.1 | - | 5 | 396 | |||

CDKL5
Genomic Map