siRNA Details
Gene Symbol: CDKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282236.1 | NP_001269165.1 | 4 | 970 | ||
NM_004196.4 | NP_004187.2 | 3 | 378 | |||
XM_005268157.2 | XP_005268214.1 | 4 | 970 | |||
XM_005268159.2 | XP_005268216.1 | 4 | 970 | |||
XM_011537276.1 | XP_011535578.1 | 4 | 380 | |||
XM_017021729.1 | XP_016877218.1 | 4 | 862 | |||
XM_017021730.1 | XP_016877219.1 | 4 | 862 | |||
XM_017021731.1 | XP_016877220.1 | 4 | 871 | |||
XM_017021732.1 | XP_016877221.1 | 4 | 862 | |||
GenBank mRNA | AK308732.1 | - | 4 | 965 | ||
BC104977.1 | - | 3 | 353 | 8143980 | ||
X66358.1 | - | 3 | 538 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
