siRNA Details
| Gene Symbol: CCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004701.3 | NP_004692.1 | 7 | 1068 | ||
| GenBank mRNA | AB020981.1 | - | 7 | 953 | ||
| AF002822.1 | - | 7 | 903 | |||
| AF087910.1 | - | 7 | 1010 | |||
| AK001404.1 | - | 7 | 1032 | |||
| AK222611.1 | - | 7 | 1001 | |||
| AL080146.1 | - | 7 | 1021 | |||
| AM851054.1 | - | 7 | 884 | |||
| BC105086.1 | - | 7 | 1016 | 8144089 | ||
| BC105112.1 | - | 7 | 1016 | 8144115 | ||
| BT019458.1 | - | 7 | 884 | |||
| CR533527.1 | - | 7 | 884 | |||

CCNB2
Genomic Map