siRNA Details
| Gene Symbol: CCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004701.3 | NP_004692.1 | 9 | 1536 | ||
| GenBank mRNA | AB020981.1 | - | 9 | 1421 | ||
| AF002822.1 | - | 9 | 1371 | |||
| AF087910.1 | - | 9 | 1478 | |||
| AK222611.1 | - | 9 | 1469 | |||

CCNB2
Genomic Map