siRNA Details
| Gene Symbol: CDK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300960.1 | NP_001287889.1 | 10 | 1122 | ||
| NM_001300963.1 | NP_001287892.1 | 12 | 1070 | |||
| NM_001300964.1 | NP_001287893.1 | 11 | 1024 | |||
| NM_015076.4 | NP_055891.1 | 11 | 1254 | |||
| XM_005266871.3 | XP_005266928.1 | 8 | 837 | |||
| XM_011535630.2 | XP_011533932.1 | 11 | 1600 | |||
| XM_011535631.2 | XP_011533933.1 | 10 | 1472 | |||
| XM_017010587.1 | XP_016866076.1 | 12 | 1480 | |||
| XM_017010588.1 | XP_016866077.1 | 9 | 946 | |||
| GenBank mRNA | AB028951.2 | - | 11 | 1071 | ||
| AK292360.1 | - | 11 | 1162 | |||
| AK300567.1 | - | 8 | 746 | |||
| AL122055.1 | - | 8 | 709 | |||
| AY028424.1 | - | 11 | 1254 | |||
| BC024247.1 | - | 12 | 1075 | |||
| BC037289.1 | - | 11 | 1149 | 5266531 | ||
| JX028238.1 | - | 10 | 942 | |||

CDK19
Genomic Map