siRNA Details
| Gene Symbol: TRIM58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015431.3 | NP_056246.3 | 1, 2 | 469 | ||
| GenBank mRNA | AF327057.1 | - | 1, 2 | 469 | ||
| AK096188.1 | - | 1, 2 | 351 | |||
| BC074748.2 | - | 13 | ||||
| BX644877.1 | - | 1, 2 | 25 | |||
| DN998591.1 | - | 1, 2 | 468 | |||

TRIM58
Genomic Map