siRNA Details
| Gene Symbol: TRIM58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015431.3 | NP_056246.3 | 5 | 901 | ||
| GenBank mRNA | AF327057.1 | - | 5 | 901 | ||
| AK096188.1 | - | 5 | 783 | |||
| BC074748.2 | - | 4 | 445 | |||
| BX537432.1 | - | 1 | 71 | |||
| BX644877.1 | - | 5 | 457 | |||

TRIM58
Genomic Map