siRNA Details
| Gene Symbol: FSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024333.2 | NP_077309.1 | 3 | 341 | ||
| XM_005259644.2 | XP_005259701.1 | 3 | 340 | |||
| GenBank mRNA | AF316829.1 | - | 3 | 232 | ||
| AK021750.1 | - | 3 | 241 | |||
| AK295523.1 | - | 3 | 236 | |||
| AK303606.1 | - | 3 | 225 | |||
| AK315661.1 | - | 3 | 335 | |||
| AY032617.1 | - | 3 | 264 | |||
| BC003124.1 | - | 3 | 241 | 3502614 | ||

FSD1
Genomic Map