siRNA Details
| Gene Symbol: CA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001218.4 | NP_001209.1 | 1, 2 | 482 | ||
| NM_001293642.1 | NP_001280571.1 | 1, 2 | 482 | |||
| NM_206925.2 | NP_996808.1 | 1, 2 | 482 | |||
| NR_135511.1 | 1, 2 | 538 | ||||
| GenBank mRNA | AF037335.1 | - | 1, 2 | 207 | ||
| AF051882.1 | - | 1, 2 | 99 | |||
| AK096845.1 | - | 1, 2 | 218 | |||
| AK315769.1 | - | 1, 2 | 248 | |||
| BC000278.1 | - | 1, 2 | 204 | 3357439 | ||
| BC011691.2 | - | 1, 2 | 207 | 3344734 | ||
| BC023981.1 | - | 1, 2 | 222 | 4042589 | ||
| BQ440047.1 | - | 1, 2 | 466 | |||
| BT006656.1 | - | 1, 2 | 92 | |||
| DA509400.1 | - | 1, 2 | 482 | |||
| DA546357.1 | - | 1, 2 | 417 | |||

CA12
Genomic Map