siRNA Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082533.1 | NP_001076002.1 | 5 | 1453 | ||
| NM_001082534.1 | NP_001076003.1 | 5 | 1018 | |||
| NM_020178.4 | NP_064563.1 | 4 | 1327 | |||
| XM_017024878.1 | XP_016880367.1 | 2 | 181 | |||
| GenBank mRNA | AB001597.1 | - | 4 | 341 | ||
| AB036836.1 | - | 5 | 1243 | |||
| AF288385.1 | - | 5 | 1009 | |||
| AK294652.1 | - | 3 | 698 | |||
| AK313001.1 | - | 4 | 903 | |||
| AL161812.1 | - | 4 | 343 | |||
| AY358509.1 | - | 4 | 621 | |||
| BC020577.1 | - | 5 | 985 | 4181377 | ||
| BC029865.1 | - | 5 | 987 | |||
| BC047456.1 | - | 4 | 1240 | 5276935 | ||
| BC068462.1 | - | 4 | 1286 | 5260321 | ||
| CR749607.1 | - | 5 | 789 | |||

CA10
Genomic Map