siRNA Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082533.1 | NP_001076002.1 | 6 | 1606 | ||
| NM_001082534.1 | NP_001076003.1 | 6 | 1171 | |||
| NM_020178.4 | NP_064563.1 | 5 | 1480 | |||
| XM_017024878.1 | XP_016880367.1 | 3 | 334 | |||
| GenBank mRNA | AB001597.1 | - | 5 | 494 | ||
| AB036836.1 | - | 6 | 1396 | |||
| AF288385.1 | - | 6 | 1162 | |||
| AK294652.1 | - | 4 | 851 | |||
| AK313001.1 | - | 5 | 1056 | |||
| AL161812.1 | - | 5 | 496 | |||
| AY358509.1 | - | 5 | 774 | |||
| BC020577.1 | - | 6 | 1138 | 4181377 | ||
| BC029865.1 | - | 6 | 1140 | |||
| BC047456.1 | - | 5 | 1393 | 5276935 | ||
| BC068462.1 | - | 5 | 1439 | 5260321 | ||
| CR749607.1 | - | 6 | 942 | |||

CA10
Genomic Map